5,5'-Dibromo-2,2'-dichloro-4,4'-bipyridine 95%
|
|
BLDPBD01578068-1GEA
176
EUR
BLDPBD01578068-1G
BLDPBD01578068-250
BLDPBD01578068-100
5,5'-Dibromo-2,2'-dichloro-4,4'-bipyridine 95%
5,5'-Dibromo-2,2'-dichloro-4,4'-bipyridine
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₀H₄Br₂Cl₂N₂ MW: 382,86 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD30178217 Núm. CAS: 1227402-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...