(7-Chloroquinolin-3-yl)boronic acid 95%

BLDPBD01542402-100EA 260 EUR
BLDPBD01542402-100 BLDPBD01542402-1G BLDPBD01542402-250
(7-Chloroquinolin-3-yl)boronic acid 95%
(7-Chloroquinolin-3-yl)boronic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₇BClNO₂
MW: 207,4214 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD22394306
Núm. CAS: 1370040-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR