(R)-4-Iodo-1-(pyrrolidin-3-yl)-1H-pyrazole hydrochloride 95%

BLDPBD01540069-100EA 138 EUR
BLDPBD01540069-100 BLDPBD01540069-250 BLDPBD01540069-1G
(R)-4-Iodo-1-(pyrrolidin-3-yl)-1H-pyrazole hydrochloride 95%
(R)-4-Iodo-1-(pyrrolidin-3-yl)-1H-pyrazole hydrochloride

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₁₁ClIN₃
MW: 299,5398 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2828444-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR