6-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)-N-(4-(6-methyl-1,2,4,5-tetrazin-3-yl)benzyl)hexanamide 95%

BLDPBD01530159-100EA 571 EUR
BLDPBD01530159-100 BLDPBD01530159-50M BLDPBD01530159-250
6-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)-N-(4-(6-methyl-1,2,4,5-tetrazin-3-yl)benzyl)hexanamide 95%
6-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)-N-(4-(6-methyl-1,2,4,5-tetrazin-3-yl)benzyl)hexanamide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₂₉H₂₉N₇O₆
MW: 571,5839 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2087490-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR