8-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline 95%

BLDPBD01528570-250EA 49 EUR
BLDPBD01528570-250 BLDPBD01528570-1G
8-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline 95%
8-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₅H₁₇BFNO₂
MW: 273,1104 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD32706315
Núm. CAS: 2235386-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR