2,6-Dimethoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 97%

BLDPBD01527026-1GEA 45 EUR
BLDPBD01527026-1G BLDPBD01527026-5G
2,6-Dimethoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 97%
2,6-Dimethoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₅H₂₁BO₅
MW: 292,13 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 2369068-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR