1-(2-Bromo-4-fluorophenyl)-N-methylmethanamine 95%

BLDPBD01517765-1GEA 176 EUR
BLDPBD01517765-1G BLDPBD01517765-250 BLDPBD01517765-100
1-(2-Bromo-4-fluorophenyl)-N-methylmethanamine 95%
1-(2-Bromo-4-fluorophenyl)-N-methylmethanamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₈H₉BrFN
MW: 218,0662 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD09810985
Núm. CAS: 887139-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR