Ethyl 4-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 97+%

BLDPBD01517343-100EA 49 EUR
BLDPBD01517343-100 BLDPBD01517343-250 BLDPBD01517343-1G
Ethyl 4-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate 97+%
Ethyl 4-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₅H₂₀BClO₄
MW: 310,58 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD28148050
Núm. CAS: 1523239-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR