4-((4-Bromophenyl)sulphonyl)-1,2-difluorobenzene 95%

BLDPBD01511625-100EA 96 EUR
BLDPBD01511625-100 BLDPBD01511625-250 BLDPBD01511625-1G
4-((4-Bromophenyl)sulphonyl)-1,2-difluorobenzene 95%
4-((4-Bromophenyl)sulphonyl)-1,2-difluorobenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₇BrF₂O₂S
MW: 333,1486 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2803456-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR