2-Amino-5-methyl-N-phenylbenzamide 95%

BLDPBD01510288-1GEA 225 EUR
BLDPBD01510288-1G BLDPBD01510288-250
2-Amino-5-methyl-N-phenylbenzamide 95%
2-Amino-5-methyl-N-phenylbenzamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₄H₁₄N₂O
MW: 226,27 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 339553-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR