1-((4-Bromophenyl)sulphonyl)-3,5-difluorobenzene 95%

BLDPBD01507080-100EA 110 EUR
BLDPBD01507080-100 BLDPBD01507080-1G BLDPBD01507080-250 BLDPBD01507080-5G
1-((4-Bromophenyl)sulphonyl)-3,5-difluorobenzene 95%
1-((4-Bromophenyl)sulphonyl)-3,5-difluorobenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₇BrF₂O₂S
MW: 333,1486 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 1708124-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR