4-(2-(2-Hydroxybenzylidene)hydrazineyl)benzoic acid 95%

BLDPBD01499845-250EA 374 EUR
BLDPBD01499845-250 BLDPBD01499845-100 BLDPBD01499845-1G
4-(2-(2-Hydroxybenzylidene)hydrazineyl)benzoic acid 95%
4-(2-(2-Hydroxybenzylidene)hydrazineyl)benzoic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₄H₁₂N₂O₃
MW: 256,25 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 325777-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR