N,N-Diisopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 95%

BLDPBD01499032-250EA 17 EUR
BLDPBD01499032-250 BLDPBD01499032-1G
N,N-Diisopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 95%
N,N-Diisopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₉H₃₀BNO₃
MW: 331,25 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 1509963-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR