tert-Butyl 4-(4-chlorobenzylidene)piperidine-1-carboxylate 95%

BLDPBD01494344-250EA 255 EUR
BLDPBD01494344-250 BLDPBD01494344-1G BLDPBD01494344-100
tert-Butyl 4-(4-chlorobenzylidene)piperidine-1-carboxylate 95%
tert-Butyl 4-(4-chlorobenzylidene)piperidine-1-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₇H₂₂ClNO₂
MW: 307,81 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD24465858
Núm. CAS: 251107-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR