(R)-4,4-Difluoro-2-methylpyrrolidine hydrochloride 95%

BLDPBD01492715-1GEA 1015 EUR
BLDPBD01492715-1G BLDPBD01492715-250 BLDPBD01492715-100
(R)-4,4-Difluoro-2-methylpyrrolidine hydrochloride 95%
(R)-4,4-Difluoro-2-methylpyrrolidine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₅H₁₀ClF₂N
MW: 157,5894 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2845095-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR