4-(4-(Trifluoromethyl)phenyl)quinoline 95%

BLDPBD01490680-100EA 58 EUR
BLDPBD01490680-100 BLDPBD01490680-250 BLDPBD01490680-1G
4-(4-(Trifluoromethyl)phenyl)quinoline 95%
4-(4-(Trifluoromethyl)phenyl)quinoline

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₆H₁₀F₃N
MW: 273,25 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2230245-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR