3-((4-Bromo-1H-pyrazol-1-yl)methyl)-5-chloropyridine 95%

BLDPBD01484130-100EA 160 EUR
BLDPBD01484130-100 BLDPBD01484130-250 BLDPBD01484130-1G
3-((4-Bromo-1H-pyrazol-1-yl)methyl)-5-chloropyridine 95%
3-((4-Bromo-1H-pyrazol-1-yl)methyl)-5-chloropyridine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₇BrClN₃
MW: 272,5290 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD30648381
Núm. CAS: 2161578-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR