tert-Butyl (S)-4-(((tert-butyldimethylsilyl)oxy)methyl)-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide 95%

BLDPBD01481990-250EA 310 EUR
BLDPBD01481990-250 BLDPBD01481990-1G BLDPBD01481990-100
tert-Butyl (S)-4-(((tert-butyldimethylsilyl)oxy)methyl)-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide 95%
tert-Butyl (S)-4-(((tert-butyldimethylsilyl)oxy)methyl)-1,2,3-oxathiazolidine-3-carboxylate 2,2-dioxide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₄H₂₉NO₆SSi
MW: 367,5337 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD30718073
Núm. CAS: 1338090-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR