Dimethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalate 95+%

BLDPBD01481651-100EA 39 EUR
BLDPBD01481651-100 BLDPBD01481651-1G BLDPBD01481651-250
Dimethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalate 95+%
Dimethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₆H₂₁BO₆
MW: 320,1454 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD23379495
Núm. CAS: 1186376-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR