cis-2,3-Dimethylazetidine hydrochloride 97%

BLDPBD01480545-1GEA 483 EUR
BLDPBD01480545-1G BLDPBD01480545-100 BLDPBD01480545-250
cis-2,3-Dimethylazetidine hydrochloride 97%
cis-2,3-Dimethylazetidine hydrochloride

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₅H₁₂ClN
MW: 121,6085 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2828444-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR