4-(4-Nitrophenyl)quinoline 95%

BLDPBD01478010-100EA 58 EUR
BLDPBD01478010-100 BLDPBD01478010-1G BLDPBD01478010-250
4-(4-Nitrophenyl)quinoline 95%
4-(4-Nitrophenyl)quinoline

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₅H₁₀N₂O₂
MW: 250,25 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 1447234-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR