(S)-5-Methyl-2,3-dihydro-1H-inden-1-amine hydrochloride 95% (by HPLC)

BLDPBD01475483-100EA 224 EUR
BLDPBD01475483-100 BLDPBD01475483-250 BLDPBD01475483-1G
(S)-5-Methyl-2,3-dihydro-1H-inden-1-amine hydrochloride 95% (by HPLC)
(S)-5-Methyl-2,3-dihydro-1H-inden-1-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₀H₁₄ClN
MW: 183,6779 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2322935-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR