1-(3-Chloro-5-methoxyphenyl)ethan-1-amine hydrochloride 95% (by HPLC)

BLDPBD01472846-1GEA 1024 EUR
BLDPBD01472846-1G BLDPBD01472846-100 BLDPBD01472846-250
1-(3-Chloro-5-methoxyphenyl)ethan-1-amine hydrochloride 95% (by HPLC)
1-(3-Chloro-5-methoxyphenyl)ethan-1-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₁₃Cl₂NO
MW: 222,1116 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD34566946
Núm. CAS: 2828440-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR