(3-Fluoro-2-methoxy-4-methylphenyl)boronic acid 95%

BLDPBD01471414-100EA 213 EUR
BLDPBD01471414-100 BLDPBD01471414-250 BLDPBD01471414-1G
(3-Fluoro-2-methoxy-4-methylphenyl)boronic acid 95%
(3-Fluoro-2-methoxy-4-methylphenyl)boronic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₁₀BFO₃
MW: 183,9726 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD22056747
Núm. CAS: 1239591-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR