Oct-3-ynoic acid 95%

BLDPBD01469463-100EA 169 EUR
BLDPBD01469463-100 BLDPBD01469463-250 BLDPBD01469463-1G
Oct-3-ynoic acid 95%
Oct-3-ynoic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₈H₁₂O₂
MW: 140,1797 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD19228277
Núm. CAS: 57074-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR