3-Fluoro-2-methylaniline hydrochloride 95%

BLDPBD01461130-5GEA 590 EUR
BLDPBD01461130-5G BLDPBD01461130-250 BLDPBD01461130-1G
3-Fluoro-2-methylaniline hydrochloride 95%
3-Fluoro-2-methylaniline hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₉ClFN
MW: 161,60 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD10698764
Núm. CAS: 654-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR