tert-Butyldiphenylsilanol 98%

BLDPBD01460716-1GEA 46 EUR
BLDPBD01460716-1G BLDPBD01460716-5G BLDPBD01460716-25G
tert-Butyldiphenylsilanol 98%
tert-Butyldiphenylsilanol

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₆H₂₀OSi
MW: 256,41 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD30533876
Núm. CAS: 93547-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR