(R)-5-Benzyl-2-(perfluorophenyl)-5,6-dihydro-8H-[1,2,4]triazolo[3,4-c][1,4]oxazin-2-ium tetrafluoroborate 97%

BLDPBD01458531-100EA 146 EUR
BLDPBD01458531-100 BLDPBD01458531-250 BLDPBD01458531-1G
(R)-5-Benzyl-2-(perfluorophenyl)-5,6-dihydro-8H-[1,2,4]triazolo[3,4-c][1,4]oxazin-2-ium tetrafluoroborate 97%
(R)-5-Benzyl-2-(perfluorophenyl)-5,6-dihydro-8H-[1,2,4]triazolo[3,4-c][1,4]oxazin-2-ium tetrafluoroborate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₈H₁₃BF₉N₃O
MW: 469,11 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 2758767-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR