(S)-1-(2-(Trifluoromethyl)pyridin-3-yl)ethan-1-amine dihydrochloride 95% (by HPLC)

BLDPBD01455284-50MEA 316 EUR
BLDPBD01455284-50M BLDPBD01455284-250 BLDPBD01455284-100
(S)-1-(2-(Trifluoromethyl)pyridin-3-yl)ethan-1-amine dihydrochloride 95% (by HPLC)
(S)-1-(2-(Trifluoromethyl)pyridin-3-yl)ethan-1-amine dihydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₁₁Cl₂F₃N₂
MW: 263,0876 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2828439-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR