5-Nitrobenzo[d][1,2,3]thiadiazole 95%

BLDPBD01454896-100EA 178 EUR
BLDPBD01454896-100 BLDPBD01454896-1G BLDPBD01454896-250 BLDPBD01454896-50M
5-Nitrobenzo[d][1,2,3]thiadiazole 95%
5-Nitrobenzo[d][1,2,3]thiadiazole

Storage condition: Sealed in dry, 2-8°C

Formula: C₆H₃N₃O₂S
MW: 181,17 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD02667407
Núm. CAS: 13599-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR