2-(2-Fluoro-3-isobutoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

BLDPBD01454530-250EA 66 EUR
BLDPBD01454530-250 BLDPBD01454530-100 BLDPBD01454530-1G
2-(2-Fluoro-3-isobutoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(2-Fluoro-3-isobutoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₆H₂₄BFO₃
MW: 294,16 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2828440-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR