(+)-tert-Butyl (4,5,6,7-tetrahydro-2H-indazol-5-yl)carbamate 98%

BLDPBD01453835-250EA 420 EUR
BLDPBD01453835-250 BLDPBD01453835-1G BLDPBD01453835-100
(+)-tert-Butyl (4,5,6,7-tetrahydro-2H-indazol-5-yl)carbamate 98%
(+)-tert-Butyl (4,5,6,7-tetrahydro-2H-indazol-5-yl)carbamate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₂H₁₉N₃O₂
MW: 237,29 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 955406-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR