(3-Bromo-4,5-dimethylphenyl)boronic acid 95+%

BLDPBD01453828-250EA 66 EUR
BLDPBD01453828-250 BLDPBD01453828-100
(3-Bromo-4,5-dimethylphenyl)boronic acid 95+%
(3-Bromo-4,5-dimethylphenyl)boronic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₁₀BBrO₂
MW: 228,8788 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 2828439-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR