(6-Hydrazineylpyridin-3-yl)(morpholino)methanone 95%

BLDPBD01453409-250EA 207 EUR
BLDPBD01453409-250 BLDPBD01453409-1G
(6-Hydrazineylpyridin-3-yl)(morpholino)methanone 95%
(6-Hydrazineylpyridin-3-yl)(morpholino)methanone

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₀H₁₄N₄O₂
MW: 222,24 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD16726478
Núm. CAS: 700803-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR