15,16-Dibromo-2,3,5,6,8,9,11,12-octahydrobenzo[b][1,4,7,10,13]pentaoxacyclopentadecine 95%

BLDPBD01452845-100EA 66 EUR
BLDPBD01452845-100 BLDPBD01452845-250 BLDPBD01452845-1G
15,16-Dibromo-2,3,5,6,8,9,11,12-octahydrobenzo[b][1,4,7,10,13]pentaoxacyclopentadecine 95%
15,16-Dibromo-2,3,5,6,8,9,11,12-octahydrobenzo[b][1,4,7,10,13]pentaoxacyclopentadecine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₄H₁₈Br₂O₅
MW: 426,09 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 63862-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR