Allyl 2-(cyclohexyloxy)acetate 95%

BLDPBD01451891-5GEA 144 EUR
BLDPBD01451891-5G BLDPBD01451891-250 BLDPBD01451891-1G
Allyl 2-(cyclohexyloxy)acetate 95%
Allyl 2-(cyclohexyloxy)acetate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₁H₁₈O₃
MW: 198,2588 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00083564
Núm. CAS: 68901-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR