2,2'-((1E,1'E)-(((1R,2R)-Cyclohexane-1,2-diyl)bis(azanylylidene))bis(methanylylidene))bis(4-(tert-butyl)phenol) 97%

BLDPBD01450989-250EA 33 EUR
BLDPBD01450989-250 BLDPBD01450989-1G BLDPBD01450989-5G
2,2'-((1E,1'E)-(((1R,2R)-Cyclohexane-1,2-diyl)bis(azanylylidene))bis(methanylylidene))bis(4-(tert-butyl)phenol) 97%
2,2'-((1E,1'E)-(((1R,2R)-Cyclohexane-1,2-diyl)bis(azanylylidene))bis(methanylylidene))bis(4-(tert-butyl)phenol)

Storage condition: Inert atmosphere, 2-8°C

Temperatura de almacenaje: Nevera
Núm. CAS: 299187-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR