(R)-2-((tert-Butoxycarbonyl)amino)-4,4,4-trifluorobutanoic acid 98%

BLDPBD01450563-250EA 318 EUR
BLDPBD01450563-250 BLDPBD01450563-100 BLDPBD01450563-1G
(R)-2-((tert-Butoxycarbonyl)amino)-4,4,4-trifluorobutanoic acid 98%
(R)-2-((tert-Butoxycarbonyl)amino)-4,4,4-trifluorobutanoic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₉H₁₄F₃NO₄
MW: 257,2070 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD09998180
Núm. CAS: 207118-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR