Ethyl (S)-1-((1-((4-((((4-nitrophenoxy)carbonyl)oxy)methyl)phenyl)amino)-1-oxo-5-ureidopentan-2-yl)carbamoyl)cyclobutane-1-carboxylate 95%

BLDPBD01449702-100EA 299 EUR
BLDPBD01449702-100 BLDPBD01449702-50M BLDPBD01449702-250 BLDPBD01449702-1G
Ethyl (S)-1-((1-((4-((((4-nitrophenoxy)carbonyl)oxy)methyl)phenyl)amino)-1-oxo-5-ureidopentan-2-yl)carbamoyl)cyclobutane-1-carboxylate 95%
Ethyl (S)-1-((1-((4-((((4-nitrophenoxy)carbonyl)oxy)methyl)phenyl)amino)-1-oxo-5-ureidopentan-2-yl)carbamoyl)cyclobutane-1-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₈H₃₃N₅O₁₀
MW: 599,58 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 2285373-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR