5-Bromo-2-((2-chlorobenzyl)oxy)-1,3-dimethylbenzene 95%

BLDPBD01449467-250EA 66 EUR
BLDPBD01449467-250 BLDPBD01449467-1G BLDPBD01449467-100
5-Bromo-2-((2-chlorobenzyl)oxy)-1,3-dimethylbenzene 95%
5-Bromo-2-((2-chlorobenzyl)oxy)-1,3-dimethylbenzene

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₅H₁₄BrClO
MW: 325,62 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD32065075
Núm. CAS: 2469394-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR