((4-(3-(3-Fluorophenyl)-5,5-dimethyl-4-oxo-4,5-dihydrofuran-2-yl)phenyl)sulphonyl)methyl acetate 95%

BLDPBD01448743-250EA 128 EUR
BLDPBD01448743-250 BLDPBD01448743-1G BLDPBD01448743-100
((4-(3-(3-Fluorophenyl)-5,5-dimethyl-4-oxo-4,5-dihydrofuran-2-yl)phenyl)sulphonyl)methyl acetate 95%
((4-(3-(3-Fluorophenyl)-5,5-dimethyl-4-oxo-4,5-dihydrofuran-2-yl)phenyl)sulphonyl)methyl acetate

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₁H₁₉FO₆S
MW: 418,4354 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD32852037
Núm. CAS: 1778703-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR