N-(Naphthalen-1-yl)-N-phenylnitrous amide 98%

BLDPBD01448715-100EA 182 EUR
BLDPBD01448715-100 BLDPBD01448715-1G BLDPBD01448715-250
N-(Naphthalen-1-yl)-N-phenylnitrous amide 98%
N-(Naphthalen-1-yl)-N-phenylnitrous amide

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₆H₁₂N₂O
MW: 248,2793 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 6341-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR