(4-(Ethoxycarbonyl)phenyl)(mesityl)iodonium tetrafluoroborate 97%

BLDPBD01448175-1GEA 81 EUR
BLDPBD01448175-1G BLDPBD01448175-5G BLDPBD01448175-250
(4-(Ethoxycarbonyl)phenyl)(mesityl)iodonium tetrafluoroborate 97%
(4-(Ethoxycarbonyl)phenyl)(mesityl)iodonium tetrafluoroborate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₈H₂₀BF₄IO₂
MW: 482,0593 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 1416353-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR