(2E,2'E)-N,N'-(1,4-Phenylenebis(methylene))bis(3-(pyridin-4-yl)acrylamide) 95%

BLDPBD01448160-1GEA 431 EUR
BLDPBD01448160-1G BLDPBD01448160-250 BLDPBD01448160-100
(2E,2'E)-N,N'-(1,4-Phenylenebis(methylene))bis(3-(pyridin-4-yl)acrylamide) 95%
(2E,2'E)-N,N'-(1,4-Phenylenebis(methylene))bis(3-(pyridin-4-yl)acrylamide)

Storage condition: Sealed in dry, Room Temperature

Formula: C₂₄H₂₂N₄O₂
MW: 398,4571 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2828433-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR