(S)-2-Chloro-5-(piperidin-2-yl)pyridine dihydrochloride 95% (by HPLC)

BLDPBD01448055-1GEA 1430 EUR
BLDPBD01448055-1G BLDPBD01448055-250 BLDPBD01448055-100
(S)-2-Chloro-5-(piperidin-2-yl)pyridine dihydrochloride 95% (by HPLC)
(S)-2-Chloro-5-(piperidin-2-yl)pyridine dihydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₀H₁₅Cl₃N₂
MW: 269,59 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2829289-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR