2-(3-Amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetic acid hydrochloride 98%

BLDPBD01447950-100EA 267 EUR
BLDPBD01447950-100 BLDPBD01447950-1G BLDPBD01447950-250
2-(3-Amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetic acid hydrochloride 98%
2-(3-Amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetic acid hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₂H₁₅ClN₂O₃
MW: 270,71 g/mol
Temperatura de almacenaje: Nevera
Núm. CAS: 2803445-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR