7-Fluoro-5-methoxyquinolin-3-amine 98%

BLDPBD01447424-100EA 220 EUR
BLDPBD01447424-100 BLDPBD01447424-1G BLDPBD01447424-250
7-Fluoro-5-methoxyquinolin-3-amine 98%
7-Fluoro-5-methoxyquinolin-3-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₀H₉FN₂O
MW: 192,1897 g/mol
Temperatura de almacenaje: Ambiente
Núm. CAS: 2828438-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR