(4S,5S)-1,3-Bis(2,6-dimethylphenyl)-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate 97%

BLDPBD01447147-250EA 339 EUR
BLDPBD01447147-250 BLDPBD01447147-1G BLDPBD01447147-100
(4S,5S)-1,3-Bis(2,6-dimethylphenyl)-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate 97%
(4S,5S)-1,3-Bis(2,6-dimethylphenyl)-4,5-diphenyl-4,5-dihydro-1H-imidazol-3-ium tetrafluoroborate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Temperatura de almacenaje: Nevera
Núm. CAS: 1033618-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR