2-(3-Chloro-4-(methoxycarbonyl)phenyl)acetic acid 95%

BLDPBD01447080-250EA 367 EUR
BLDPBD01447080-250 BLDPBD01447080-1G BLDPBD01447080-100
2-(3-Chloro-4-(methoxycarbonyl)phenyl)acetic acid 95%
2-(3-Chloro-4-(methoxycarbonyl)phenyl)acetic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₉ClO₄
MW: 228,6291 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD32185722
Núm. CAS: 1324054-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR