2,5-Dioxopyrrolidin-1-yl 3-iodobenzoate 98%

BLDPBD01445777-5GEA 881 EUR
BLDPBD01445777-5G BLDPBD01445777-100 BLDPBD01445777-1G
2,5-Dioxopyrrolidin-1-yl 3-iodobenzoate 98%
2,5-Dioxopyrrolidin-1-yl 3-iodobenzoate

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₁H₈INO₄
MW: 345,0900 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD27964177
Núm. CAS: 91487-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR